siRNA Details
Gene Symbol: DNM1P46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003260.1 | 5 | 1508 | |||
GenBank mRNA | AK127834.1 | - | 5 | 1508 | ||
AK302717.1 | - | 5 | 1309 | |||
AL833912.1 | - | 4 | 1363 |
Gene Symbol: LOC440300 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033738.1 | 8 | 2903 | |||
GenBank mRNA | BX648930.1 | - | 8 | 2903 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
