siRNA Details
Gene Symbol: CKS1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001826.2 | NP_001817.1 | 2 | 216 | ||
NR_024163.1 | 2 | 324 | ||||
GenBank mRNA | AF274941.1 | - | 1 | 138 | ||
AF274943.1 | - | 2 | 144 | |||
AF279897.1 | - | 2 | 138 | |||
AK307066.1 | - | 2 | 272 | |||
AK311769.1 | - | 2 | 177 | |||
BC007751.2 | - | 2 | 159 | 4110344 | ||
BC015629.2 | - | 2 | 138 | 3162564 | ||
BC070319.1 | - | 2 | 177 | 4738841 | ||
BC070320.1 | - | 2 | 177 | 6494754 | ||
BQ883992.1 | - | 2 | 155 | |||
BT007196.1 | - | 2 | 112 | |||
CB161621.1 | - | 2 | 275 | |||
CN347951.1 | - | 2 | 218 | |||
X54941.1 | - | 2 | 121 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
