siRNA Details
Gene Symbol: HNRNPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002136.3 | NP_002127.1 | 2 | 184 | ||
NM_031157.3 | NP_112420.1 | 2 | 184 | |||
NR_135167.1 | 2 | 184 | ||||
XM_005268826.1 | XP_005268883.1 | 2 | 178 | |||
XM_017019251.1 | XP_016874740.1 | 2 | 178 | |||
GenBank mRNA | AK129814.1 | - | 2 | 126 | ||
AK291113.1 | - | 2 | 124 | |||
AK298176.1 | - | 2 | 127 | |||
AK303303.1 | - | 2 | 126 | |||
AK308679.1 | - | 2 | 126 | |||
BC002355.2 | - | 2 | 105 | 2821751 | ||
BC009600.1 | - | 2 | 127 | 4134638 | ||
BC012158.1 | - | 2 | 87 | 4564655 | ||
BC020442.2 | - | 1 | 94 | |||
BC033714.1 | - | 2 | 116 | 4420039 | ||
BC052296.1 | - | 2 | 96 | 6297642 | ||
BC070315.1 | - | 2 | 126 | 4101267 | ||
BC071945.1 | - | 1 | 116 | 5475712 | ||
BC073162.1 | - | 1 | 110 | 6154972 | ||
BC074502.1 | - | 2 | 109 | 3455742 | ||
BC103707.1 | - | 2 | 94 | 6154800 | ||
BC121133.2 | - | 2 | 110 | 40120894 | ||
BQ277208.1 | - | 184 | ||||
BX425482.2 | - | 2 | 108 | |||
CD705336.1 | - | 2 | 201 | |||
DC358304.1 | - | 2 | 280 | |||
U00947.1 | - | 2 | 74 | |||
X06747.1 | - | 2 | 151 | |||
X79536.1 | - | 2 | 92 |
Gene Symbol: HNRNPA1L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011724.1 | NP_001011724.1 | 7 | 1139 | ||
NM_001011725.1 | NP_001011725.1 | 6 | 998 | |||
GenBank mRNA | AK091082.1 | - | 1 | 851 | ||
AK094423.1 | - | 6 | 889 | |||
AK123159.1 | - | 5 | 678 | |||
AK126454.1 | - | 1 | 1773 | |||
BC082242.1 | - | 6 | 929 | |||
BC108266.1 | - | 1 | 127 | 4778431 |
Gene Symbol: HNRNPA1P10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002944.2 | 1 | 125 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
