siRNA Details
Gene Symbol: DLL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016941.3 | NP_058637.1 | 8 | 1944 | ||
GenBank mRNA | BC000218.2 | - | 8 | 1893 | 3508262 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
