siRNA Details
Gene Symbol: HNRNPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002136.3 | NP_002127.1 | 4 | 514 | ||
NM_031157.3 | NP_112420.1 | 4 | 514 | |||
NR_135167.1 | 4 | 514 | ||||
XM_005268826.1 | XP_005268883.1 | 4 | 508 | |||
XM_017019251.1 | XP_016874740.1 | 4 | 508 | |||
GenBank mRNA | AK129814.1 | - | 4 | 456 | ||
AK291113.1 | - | 4 | 454 | |||
AK308679.1 | - | 3 | 408 | |||
BC002355.2 | - | 4 | 435 | 2821751 | ||
BC004945.1 | - | 1 | 331 | |||
BC009600.1 | - | 4 | 457 | 4134638 | ||
BC012158.1 | - | 4 | 417 | 4564655 | ||
BC020442.2 | - | 1 | 424 | |||
BC033714.1 | - | 4 | 446 | 4420039 | ||
BC052296.1 | - | 4 | 426 | 6297642 | ||
BC070315.1 | - | 4 | 456 | 4101267 | ||
BC071945.1 | - | 1 | 446 | 5475712 | ||
BC073162.1 | - | 1 | 440 | 6154972 | ||
BC074502.1 | - | 4 | 439 | 3455742 | ||
BC103707.1 | - | 4 | 424 | 6154800 | ||
BC121133.2 | - | 4 | 440 | 40120894 | ||
BU541100.1 | - | 1 | 84 | |||
BX425482.2 | - | 4 | 438 | |||
U00947.1 | - | 4 | 404 | |||
X04347.1 | - | 1 | 24 | |||
X06747.1 | - | 4 | 481 | |||
X79536.1 | - | 4 | 422 |
Gene Symbol: HNRNPA1P10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002944.2 | 1 | 455 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
