siRNA Details
Gene Symbol: FAM230B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108107.1 | 3, 4 | 302 | |||
GenBank mRNA | BC039313.1 | - | 3, 4 | 302 |
Gene Symbol: FAM230C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027278.1 | 3, 4 | 205 | |||
GenBank mRNA | AK097777.1 | - | 3, 4 | 151 | ||
DB056745.1 | - | 2, 3 | 82 |
Gene Symbol: LOC100996335 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136559.1 | 3, 4 | 194 | |||
GenBank mRNA | DB096903.1 | - | 3, 4 | 194 |
Gene Symbol: LOC100996432 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136571.1 | 3, 4 | 193 | |||
GenBank mRNA | DB073469.1 | - | 3, 4 | 171 |
Gene Symbol: LOC101927859 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136572.1 | 2, 3 | 90 |
Gene Symbol: LOC642643 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136561.1 | 3, 4 | 231 |
Gene Symbol: LOC729461 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136569.1 | 3, 4 | 231 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
