siRNA Details
Gene Symbol: LOC107985931 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017005472.1 | XP_016860961.1 | 3 | 228 | ||
XM_017005473.1 | XP_016860962.1 | 3 | 185 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
