siRNA Details
Gene Symbol: GH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000515.4 | NP_000506.2 | 5 | 792 | ||
NM_022559.3 | NP_072053.1 | 5 | 747 | |||
NM_022560.3 | NP_072054.1 | 4 | 672 | |||
GenBank mRNA | A00469.1 | - | 5 | 755 | ||
AF110644.1 | - | 5 | 588 | |||
AF185611.1 | - | 6 | 620 | |||
AV753220.1 | - | 3 | 612 | |||
AY613431.1 | - | 5 | 778 | |||
AY613432.1 | - | 5 | 778 | |||
BC062475.1 | - | 5 | 712 | 30393581 | ||
BC090045.1 | - | 5 | 759 | 30394115 | ||
CD108592.1 | - | 4 | 638 | |||
V00519.1 | - | 5 | 756 |

More Information
Other Species Targeted

Gene Ontology Categories:
