siRNA Details
Gene Symbol: GH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000515.4 | NP_000506.2 | 5 | 575 | ||
NM_022559.3 | NP_072053.1 | 5 | 530 | |||
NM_022560.3 | NP_072054.1 | 4 | 455 | |||
GenBank mRNA | A00469.1 | - | 5 | 538 | ||
AF110644.1 | - | 5 | 371 | |||
AF185611.1 | - | 6 | 403 | |||
AV753220.1 | - | 3 | 395 | |||
AY613431.1 | - | 5 | 561 | |||
AY613432.1 | - | 5 | 561 | |||
BC062475.1 | - | 5 | 495 | 30393581 | ||
BC075012.2 | - | 5 | 526 | 30915404 | ||
BC075013.2 | - | 5 | 526 | 30915617 | ||
BC090045.1 | - | 5 | 542 | 30394115 | ||
CD108592.1 | - | 4 | 421 | |||
V00519.1 | - | 5 | 539 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
