siRNA Details
Gene Symbol: GH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000515.4 | NP_000506.2 | 5 | 654 | ||
NM_022559.3 | NP_072053.1 | 5 | 609 | |||
NM_022560.3 | NP_072054.1 | 4 | 534 | |||
GenBank mRNA | A00469.1 | - | 5 | 617 | ||
AF110644.1 | - | 5 | 450 | |||
AF185611.1 | - | 6 | 482 | |||
AV753220.1 | - | 3 | 474 | |||
AY613431.1 | - | 5 | 640 | |||
AY613432.1 | - | 5 | 640 | |||
BC062475.1 | - | 5 | 574 | 30393581 | ||
BC075012.2 | - | 5 | 605 | 30915404 | ||
BC075013.2 | - | 5 | 605 | 30915617 | ||
BC090045.1 | - | 5 | 621 | 30394115 | ||
CD108592.1 | - | 4 | 500 | |||
V00519.1 | - | 5 | 618 |
Gene Symbol: CSH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317.5 | NP_001308.1 | 5 | 728 | ||
GenBank mRNA | AK291587.1 | - | 5 | 647 | ||
BC002717.2 | - | 5 | 621 | 3631916 | ||
BC005921.1 | - | 5 | 640 | 4043768 | ||
BC020756.1 | - | 5 | 696 | 4769769 | ||
BC057768.1 | - | 5 | 640 | 30337725 | ||
BC062775.1 | - | 5 | 644 | 4604848 | ||
BT006926.1 | - | 4 | 578 | |||
J00118.1 | - | 5 | 607 | |||
M25118.1 | - | 1 | 101 | |||
V00593.1 | - | 4 | 430 |
Gene Symbol: CSH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020991.3 | NP_066271.1 | 5 | 693 | ||
NM_022644.3 | NP_072170.1 | 4 | 946 | |||
NM_022645.2 | NP_072171.1 | 3 | 408 | |||
GenBank mRNA | BC022044.1 | - | 5 | 644 | 4769046 | |
BC035965.1 | - | 5 | 640 | 4731880 | ||
BC119748.2 | - | 5 | 637 | 40119242 | ||
V00573.1 | - | 5 | 602 |
Gene Symbol: GH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002059.4 | NP_002050.1 | 5 | 719 | ||
NM_022556.3 | NP_072050.1 | 5 | 674 | |||
NM_022557.3 | NP_072051.1 | 4 | 972 | |||
NM_022558.3 | NP_072052.1 | 5 | 715 | |||
GenBank mRNA | AB451332.1 | - | 4 | 533 | ||
AB451477.1 | - | 4 | 533 | |||
AF006060.1 | - | 5 | 545 | |||
AF006061.1 | - | 4 | 574 | |||
BC020760.1 | - | 5 | 640 | 4720475 | ||
CR541902.1 | - | 4 | 578 | |||
CR541924.1 | - | 4 | 578 | |||
J03756.1 | - | 4 | 892 | |||
M38451.1 | - | 5 | 590 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
