siRNA Details
Gene Symbol: LINC00266-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040415.1 | 2 | 443 | |||
GenBank mRNA | BC118988.1 | - | 2 | 443 | 40035995 | |
BC122537.1 | - | 2 | 443 | 40035996 |
Gene Symbol: LINC00266-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109817.1 | 3 | 419 | |||
GenBank mRNA | DB459734.1 | - | 3 | 421 |
Gene Symbol: LOC100134822 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL137655.1 | - | 17 | 3085 |
Gene Symbol: LOC101927506 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017014131.1 | XP_016869620.1 | 2 | 2020 | ||
XM_017014132.1 | XP_016869621.1 | 2 | 2020 | |||
XM_017014133.1 | XP_016869622.1 | 2 | 2020 | |||
XM_017014134.1 | XP_016869623.1 | 2 | 2020 | |||
XM_017014135.1 | XP_016869624.1 | 2 | 2020 | |||
XM_017014136.1 | XP_016869625.1 | 2 | 2020 |
Gene Symbol: LOC728323 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130699.1 | 5 | 599 | |||
NR_130700.1 | 4 | 496 | ||||
NR_130701.1 | 6 | 649 | ||||
GenBank mRNA | AK298283.1 | - | 5 | 539 | ||
AK307199.1 | - | 6 | 589 | |||
AK309231.1 | - | 4 | 445 | |||
BC131690.1 | - | 2 | 112 |
Gene Symbol: OR4F16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017002409.1 | XP_016857898.1 | 11 |
Gene Symbol: PCMTD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC032332.1 | - | 7 | 1141 | 5169056 |
Gene Symbol: SEPT14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207366.2 | NP_997249.2 | 10 | 1329 | ||
XM_011515373.2 | XP_011513675.1 | 10 | 1313 | |||
GenBank mRNA | AK126048.1 | - | 5 | 617 | ||
AK301928.1 | - | 10 | 1292 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
