siRNA Details
Gene Symbol: SEPT14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207366.2 | NP_997249.2 | 10 | 1253 | ||
XM_011515373.2 | XP_011513675.1 | 10 | 1237 | |||
GenBank mRNA | AK126048.1 | - | 5 | 541 | ||
AK301928.1 | - | 10 | 1216 | |||
DB087561.1 | - | 5 | 541 |
Gene Symbol: LOC100134822 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL137655.1 | - | 17 | 3009 |
Gene Symbol: LOC101927506 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017014131.1 | XP_016869620.1 | 2 | 1944 | ||
XM_017014132.1 | XP_016869621.1 | 2 | 1944 | |||
XM_017014133.1 | XP_016869622.1 | 2 | 1944 | |||
XM_017014134.1 | XP_016869623.1 | 2 | 1944 | |||
XM_017014135.1 | XP_016869624.1 | 2 | 1944 | |||
XM_017014136.1 | XP_016869625.1 | 2 | 1944 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
