siRNA Details
Gene Symbol: NUTM2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099338.1 | NP_001092808.1 | 6 | 2174 | ||
XM_011540122.2 | XP_011538424.1 | 8 | 5017 | |||
XM_017016607.1 | XP_016872096.1 | 8 | 5018 | |||
GenBank mRNA | BC092519.1 | - | 6 | 2170 | ||
EG328792.1 | - | 451 |
Gene Symbol: NUTM2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278495.1 | NP_001265424.1 | 6 | 2174 |
Gene Symbol: NUTM2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_075100.1 | 6 | 2121 | |||
GenBank mRNA | AB095940.1 | - | 6 | 2121 |
Gene Symbol: NUTM2F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017561.1 | NP_060031.1 | 6 | 1446 | ||
GenBank mRNA | AL133071.1 | - | 5 | 999 | ||
BC130390.1 | - | 6 | 1446 | 40146421 | ||
BC143798.1 | - | 6 | 1446 | 9052312 |
Gene Symbol: NUTM2G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045477.2 | NP_001038942.1 | 6 | 1401 | ||
NM_001170741.1 | NP_001164212.1 | 6 | 1401 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
