siRNA Details
Gene Symbol: AGAP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144000.1 | NP_001137472.1 | 8 | 1109 |
Gene Symbol: AGAP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133447.1 | NP_597704.1 | 12 | 1789 | ||
GenBank mRNA | AB075855.1 | - | 7 | 3193 | ||
AK097165.1 | - | 12 | 1789 | |||
BC140762.1 | - | 8 | 1035 | 9021630 |
Gene Symbol: AGAP12P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029396.2 | 8 | 1110 |
Gene Symbol: AGAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF411132.1 | - | 7 | 1125 | ||
BC048341.1 | - | 1 | 247 | |||
BC171820.1 | - | 8 | 1179 |
Gene Symbol: AGAP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077665.2 | NP_001071133.2 | 8 | 1466 | ||
XM_005269841.3 | XP_005269898.1 | 2 | 507 | |||
XM_011539814.2 | XP_011538116.1 | 2 | 511 | |||
XM_011539815.2 | XP_011538117.1 | 5 | 691 | |||
XM_017016268.1 | XP_016871757.1 | 8 | 1187 | |||
XM_017016269.1 | XP_016871758.1 | 8 | 1187 | |||
XM_017016270.1 | XP_016871759.1 | 7 | 1421 | |||
XM_017016271.1 | XP_016871760.1 | 10 | 1189 | |||
XM_017016272.1 | XP_016871761.1 | 9 | 1119 | |||
XM_017016273.1 | XP_016871762.1 | 9 | 1812 | |||
XM_017016274.1 | XP_016871763.1 | 5 | 642 |
Gene Symbol: AGAP7P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126580.1 | 7 | 1094 | |||
GenBank mRNA | BY996334.1 | - | 1 | 159 |
Gene Symbol: AGAP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190810.1 | NP_001177739.1 | 8 | 1180 | ||
GenBank mRNA | AK160375.1 | - | 9 | 4156 | ||
BC171845.1 | - | 8 | 1180 |
Gene Symbol: LOC107984026 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017017029.1 | XP_016872518.1 | 10 | 1611 | ||
XM_017017030.1 | XP_016872519.1 | 9 | 1542 | |||
XM_017017031.1 | XP_016872520.1 | 9 | 1542 | |||
XM_017017032.1 | XP_016872521.1 | 11 | 1642 | |||
XM_017017033.1 | XP_016872522.1 | 12 | 1405 | |||
XM_017017034.1 | XP_016872523.1 | 11 | 1310 | |||
XM_017017035.1 | XP_016872524.1 | 7 | 823 | |||
XM_017017036.1 | XP_016872525.1 | 5 | 984 | |||
XM_017017037.1 | XP_016872526.1 | 7 | 1373 | |||
XM_017017038.1 | XP_016872527.1 | 5 | 626 | |||
XM_017017039.1 | XP_016872528.1 | 7 | 842 | |||
XM_017017040.1 | XP_016872529.1 | 7 | 806 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
