siRNA Details
Gene Symbol: AGAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276343.2 | NP_001263272.2 | 8 | 2195 | ||
NM_001291379.1 | NP_001278308.1 | 10 | 2387 | |||
NM_133446.3 | NP_597703.2 | 7 | 2126 | |||
XM_011539269.2 | XP_011537571.1 | 6 | 1733 | |||
XM_017015664.1 | XP_016871153.1 | 10 | 6057 | |||
XM_017015665.1 | XP_016871154.1 | 11 | 5925 | |||
XM_017015666.1 | XP_016871155.1 | 9 | 2960 | |||
XM_017015667.1 | XP_016871156.1 | 10 | 2230 | |||
XM_017015668.1 | XP_016871157.1 | 8 | 1980 | |||
XM_017015669.1 | XP_016871158.1 | 8 | 1888 | |||
GenBank mRNA | AF411132.1 | - | 7 | 2126 | ||
BC048341.1 | - | 1 | 1248 | |||
BC171820.1 | - | 8 | 2180 |
Gene Symbol: AGAP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133447.1 | NP_597704.1 | 12 | 2865 | ||
GenBank mRNA | AB075855.1 | - | 7 | 4269 | ||
BC140762.1 | - | 8 | 2111 | 9021630 |
Gene Symbol: AGAP12P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029396.2 | 8 | 2111 |
Gene Symbol: AGAP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144000.1 | NP_001137472.1 | 8 | 2110 |
Gene Symbol: AGAP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077665.2 | NP_001071133.2 | 8 | 2467 | ||
XM_005269841.3 | XP_005269898.1 | 2 | 1508 | |||
XM_011539814.2 | XP_011538116.1 | 2 | 1512 | |||
XM_011539815.2 | XP_011538117.1 | 5 | 1692 | |||
XM_017016268.1 | XP_016871757.1 | 8 | 2188 | |||
XM_017016269.1 | XP_016871758.1 | 8 | 2188 | |||
XM_017016270.1 | XP_016871759.1 | 7 | 2422 | |||
XM_017016271.1 | XP_016871760.1 | 10 | 2190 | |||
XM_017016272.1 | XP_016871761.1 | 9 | 2120 | |||
XM_017016273.1 | XP_016871762.1 | 9 | 2813 | |||
XM_017016274.1 | XP_016871763.1 | 5 | 1643 |
Gene Symbol: AGAP7P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126580.1 | 7 | 2095 |
Gene Symbol: AGAP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190810.1 | NP_001177739.1 | 8 | 2174 | ||
GenBank mRNA | AK160375.1 | - | 10 | 5090 | ||
BC171845.1 | - | 8 | 2174 |
Gene Symbol: LOC107984026 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017017029.1 | XP_016872518.1 | 10 | 2612 | ||
XM_017017030.1 | XP_016872519.1 | 9 | 2543 | |||
XM_017017031.1 | XP_016872520.1 | 9 | 2543 | |||
XM_017017032.1 | XP_016872521.1 | 11 | 2643 | |||
XM_017017033.1 | XP_016872522.1 | 12 | 2406 | |||
XM_017017034.1 | XP_016872523.1 | 11 | 2311 | |||
XM_017017035.1 | XP_016872524.1 | 7 | 1824 | |||
XM_017017036.1 | XP_016872525.1 | 5 | 1985 | |||
XM_017017037.1 | XP_016872526.1 | 7 | 2374 | |||
XM_017017038.1 | XP_016872527.1 | 5 | 1627 | |||
XM_017017039.1 | XP_016872528.1 | 7 | 1843 | |||
XM_017017040.1 | XP_016872529.1 | 7 | 1807 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
