siRNA Details
Gene Symbol: RPL7L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198486.3 | NP_940888.3 | 6 | 1013 | ||
NR_134562.1 | 6 | 1129 | ||||
NR_134563.1 | 5 | 907 | ||||
XM_017010780.1 | XP_016866269.1 | 6 | 1466 | |||
GenBank mRNA | AK021904.1 | - | 5 | 594 | ||
AK291310.1 | - | 5 | 616 | |||
AK291949.1 | - | 6 | 700 | |||
AK299816.1 | - | 5 | 602 | |||
BC058020.1 | - | 6 | 704 | 6736635 | ||
BC073890.1 | - | 6 | 696 | 5491531 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
