siRNA Details
Gene Symbol: MXRA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282582.1 | NP_001269511.1 | 7 | 1134 | ||
NM_001282583.1 | NP_001269512.1 | 6 | 1553 | |||
NM_001282584.1 | NP_001269513.1 | 5 | 1053 | |||
NM_001282585.1 | NP_001269514.1 | 6 | 1109 | |||
NM_032348.3 | NP_115724.1 | 6 | 1356 | |||
XM_017001516.1 | XP_016857005.1 | 6 | 1133 | |||
XM_017001517.1 | XP_016857006.1 | 5 | 2760 | |||
GenBank mRNA | AB052096.1 | - | 7 | 1134 | ||
AK027269.1 | - | 6 | 1109 | |||
AK095966.1 | - | 6 | 1553 | |||
AK225028.1 | - | 6 | 1095 | |||
AK293442.1 | - | 5 | 822 | |||
AK304615.1 | - | 6 | 1027 | |||
AK315510.1 | - | 6 | 1123 | |||
AY358391.1 | - | 5 | 799 | |||
BC006213.2 | - | 6 | 1097 | 3343261 | ||
BC017312.2 | - | 6 | 1097 | 5090754 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
