siRNA Details
Gene Symbol: MXRA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282582.1 | NP_001269511.1 | 10 | 1359 | ||
NM_001282583.1 | NP_001269512.1 | 9 | 1778 | |||
NM_001282584.1 | NP_001269513.1 | 8 | 1278 | |||
NM_001282585.1 | NP_001269514.1 | 9 | 1352 | |||
NM_032348.3 | NP_115724.1 | 9 | 1581 | |||
XM_017001516.1 | XP_016857005.1 | 9 | 1358 | |||
XM_017001517.1 | XP_016857006.1 | 8 | 2985 | |||
GenBank mRNA | AB052096.1 | - | 10 | 1359 | ||
AK027269.1 | - | 9 | 1352 | |||
AK095966.1 | - | 9 | 1778 | |||
AK225028.1 | - | 9 | 1320 | |||
AK293442.1 | - | 8 | 1047 | |||
AK304615.1 | - | 8 | 1373 | |||
AK315510.1 | - | 9 | 1348 | |||
AL050202.1 | - | 3 | 217 | |||
AY358391.1 | - | 8 | 1024 | |||
BC006213.2 | - | 9 | 1322 | 3343261 | ||
BC017312.2 | - | 9 | 1322 | 5090754 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
