siRNA Details
Gene Symbol: FAM35A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019054.2 | NP_061927.2 | 4 | 1737 | ||
XM_005269920.3 | XP_005269977.1 | 4 | 680 | |||
XM_006717903.3 | XP_006717966.1 | 5 | 1871 | |||
XM_006717905.3 | XP_006717968.1 | 4 | 1794 | |||
XM_011539876.2 | XP_011538178.1 | 3 | 1772 | |||
XM_011539877.2 | XP_011538179.1 | 6 | 1936 | |||
XM_011539878.2 | XP_011538180.1 | 4 | 1794 | |||
XM_011539879.2 | XP_011538181.1 | 3 | 1780 | |||
XM_011539881.2 | XP_011538183.1 | 3 | 1772 | |||
XM_011539882.2 | XP_011538184.1 | 3 | 1772 | |||
XM_011539883.2 | XP_011538185.1 | 3 | 725 | |||
XM_017016349.1 | XP_016871838.1 | 8 | 2180 | |||
XM_017016350.1 | XP_016871839.1 | 5 | 2139 | |||
XM_017016351.1 | XP_016871840.1 | 4 | 1727 | |||
XM_017016352.1 | XP_016871841.1 | 4 | 1727 | |||
XM_017016353.1 | XP_016871842.1 | 4 | 680 | |||
GenBank mRNA | AF131775.1 | - | 1 | 39 | ||
AK022978.1 | - | 4 | 1737 | |||
AK025606.1 | - | 2 | 586 | |||
BC051863.2 | - | 3 | 1683 | 6146593 |
Gene Symbol: FAM35BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027632.1 | 2 | 1628 |
Gene Symbol: FAM35DP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027634.1 | 2 | 1628 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
