siRNA Details
Gene Symbol: OR2T35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001827.1 | NP_001001827.1 | 1 | 577 |
Gene Symbol: OR2T2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004136.1 | NP_001004136.1 | 1 | 580 | ||
GenBank mRNA | BC136961.1 | - | 1 | 602 | 9020953 | |
BC136962.1 | - | 1 | 602 | 9020954 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
