siRNA Details
Gene Symbol: DHFR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000791.3 | NP_000782.1 | 6 | 1007 | ||
NM_001290354.1 | NP_001277283.1 | 5 | 957 | |||
NM_001290357.1 | NP_001277286.1 | 5 | 891 | |||
NR_110936.1 | 4 | 830 | ||||
GenBank mRNA | AK293146.1 | - | 5 | 535 | ||
AK297302.1 | - | 5 | 462 | |||
AK308492.1 | - | 4 | 397 | |||
AK312642.1 | - | 1 | 551 | |||
BC000192.1 | - | 6 | 570 | 3352539 | ||
BC003584.1 | - | 7 | 969 | 3354190 | ||
BC070280.1 | - | 6 | 552 | 6714220 | ||
BC071996.1 | - | 7 | 969 | 4645596 | ||
EU145592.1 | - | 1 | 515 | |||
J00140.1 | - | 6 | 557 | |||
V00507.1 | - | 1 | 515 |
Gene Symbol: LOC1720 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033423.1 | 1 | 537 | |||
GenBank mRNA | J00146.1 | - | 1 | 534 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
