siRNA Details
Gene Symbol: DHFR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000791.3 | NP_000782.1 | 3 | 705 | ||
NM_001290354.1 | NP_001277283.1 | 2 | 655 | |||
NM_001290357.1 | NP_001277286.1 | 3 | 705 | |||
NR_110936.1 | 2 | 655 | ||||
GenBank mRNA | AK293146.1 | - | 2 | 233 | ||
AK297302.1 | - | 3 | 276 | |||
AK308492.1 | - | 2 | 222 | |||
AK312642.1 | - | 1 | 249 | |||
BC000192.1 | - | 3 | 268 | 3352539 | ||
BC003584.1 | - | 4 | 667 | 3354190 | ||
BC070280.1 | - | 3 | 250 | 6714220 | ||
BC071996.1 | - | 4 | 667 | 4645596 | ||
EU145592.1 | - | 1 | 213 | |||
J00140.1 | - | 3 | 255 | |||
V00507.1 | - | 1 | 213 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
