siRNA Details
Gene Symbol: NBPF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015383.2 | NP_056198.2 | 10 | 1487 | ||
XM_011509395.2 | XP_011507697.1 | 13 | 1987 | |||
GenBank mRNA | AL117237.1 | - | 4 | 484 |
Gene Symbol: LOC102724250 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011546188.2 | XP_011544490.1 | 12 | 10403 | ||
XM_017030107.1 | XP_016885596.1 | 12 | 10403 |
Gene Symbol: LOC105371303 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003020.1 | XP_016858509.1 | 10 | 1637 |
Gene Symbol: NBPF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017940.4 | NP_060410.2 | 3306 | |||
GenBank mRNA | AB051480.1 | - | 11 | 1363 | ||
AF379623.1 | - | 2 | 1730 | |||
AF379624.1 | - | 1 | 91 | |||
AY894575.1 | - | 22 | 3188 | |||
BC034418.1 | - | 6 | 868 | |||
BC094705.1 | - | 4 | 514 |
Gene Symbol: NBPF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039703.5 | NP_001034792.4 | 1522 | |||
NM_001302371.1 | NP_001289300.1 | 1522 | ||||
GenBank mRNA | AF379635.1 | - | 6 | 785 | ||
AK000726.1 | - | 10 | 2036 | |||
BC021111.1 | - | 11 | 1566 | |||
BC063799.1 | - | 11 | 1549 |
Gene Symbol: NBPF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278141.1 | NP_001265070.1 | 21 | 3517 | ||
GenBank mRNA | AB033071.1 | - | 21 | 3070 |
Gene Symbol: NBPF19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003019.1 | XP_016858508.1 | 7 | 947 | ||
GenBank mRNA | AK295639.1 | - | 1 | 418 |
Gene Symbol: NBPF20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278267.1 | NP_001265196.1 | 806 |
Gene Symbol: NBPF25P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104217.1 | 9 | 1131 | |||
GenBank mRNA | AK125792.1 | - | 9 | 1131 |
Gene Symbol: NBPF26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011542529.2 | XP_011540831.1 | 10 | 1517 | ||
XM_017003011.1 | XP_016858500.1 | 10 | 1516 | |||
GenBank mRNA | AK294944.1 | - | 4 | 500 |
Gene Symbol: NBPF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037501.2 | NP_001032590.2 | 10 | 1659 | ||
NR_102404.1 | 10 | 1659 | ||||
NR_102405.1 | 9 | 1556 | ||||
GenBank mRNA | AK307809.1 | - | 8 | 1263 | ||
AY894561.1 | - | 6 | 848 | |||
AY894562.1 | - | 6 | 848 | |||
AY894563.1 | - | 5 | 722 | |||
AY894568.1 | - | 10 | 1454 | |||
AY894569.1 | - | 10 | 1456 | |||
AY894570.1 | - | 6 | 848 | |||
AY894572.1 | - | 10 | 1659 | |||
BC086308.1 | - | 10 | 1502 | 5453529 |
Gene Symbol: NBPF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037675.3 | NP_001032764.2 | 11 | 1562 | ||
NM_001277444.1 | NP_001264373.1 | 11 | 1562 | |||
XM_006711317.2 | XP_006711380.1 | 12 | 1790 | |||
XM_017001278.1 | XP_016856767.1 | 12 | 1790 | |||
GenBank mRNA | AF419618.1 | - | 4 | 510 | ||
AK294414.1 | - | 5 | 720 | |||
AY894574.1 | - | 11 | 1564 | |||
BC071995.1 | - | 6 | 746 | |||
BC094841.1 | - | 6 | 746 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
