siRNA Details
Gene Symbol: NBPF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101663.4 | NP_001095133.3 | 12 | 1718 | ||
NM_183372.5 | NP_899228.4 | 14 | 2412 | |||
NR_046188.3 | 15 | 2560 | ||||
NR_110864.1 | 13 | 2328 | ||||
GenBank mRNA | AF419617.1 | - | 13 | 1831 | ||
AK302413.1 | - | 11 | 1554 | |||
AL832622.2 | - | 15 | 2693 | |||
BC136292.1 | - | 14 | 2412 | 9020279 | ||
BX648497.1 | - | 13 | 2127 |
Gene Symbol: LOC102724250 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011546188.2 | XP_011544490.1 | 12 | 10309 | ||
XM_017030107.1 | XP_016885596.1 | 12 | 10309 |
Gene Symbol: LOC105371303 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003020.1 | XP_016858509.1 | 10 | 1543 |
Gene Symbol: NBPF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017940.4 | NP_060410.2 | 3212 | |||
GenBank mRNA | AY894575.1 | - | 22 | 3094 | ||
BC094705.1 | - | 4 | 420 |
Gene Symbol: NBPF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039703.5 | NP_001034792.4 | 1428 | |||
NM_001302371.1 | NP_001289300.1 | 1428 | ||||
GenBank mRNA | AF379635.1 | - | 6 | 691 | ||
AK000726.1 | - | 10 | 1942 | |||
AY894565.1 | - | 11 | 1465 | |||
BC021111.1 | - | 11 | 1472 | |||
BC063799.1 | - | 11 | 1455 |
Gene Symbol: NBPF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278141.1 | NP_001265070.1 | 21 | 3423 | ||
GenBank mRNA | AB033071.1 | - | 21 | 2976 |
Gene Symbol: NBPF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015383.2 | NP_056198.2 | 10 | 1393 | ||
XM_011509395.2 | XP_011507697.1 | 13 | 1893 | |||
GenBank mRNA | AL117237.1 | - | 4 | 390 |
Gene Symbol: NBPF15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170755.2 | NP_001164226.1 | 12 | 1742 | ||
NM_173638.4 | NP_775909.2 | 9 | 1212 | |||
XM_017001077.1 | XP_016856566.1 | 7 | 860 | |||
GenBank mRNA | AF379630.1 | - | 4 | 390 | ||
AK097180.1 | - | 12 | 1661 | |||
AK290302.1 | - | 6 | 671 | |||
AK299360.1 | - | 3 | 252 | |||
AY894566.1 | - | 6 | 652 | |||
BC023087.1 | - | 9 | 1069 | 3918932 | ||
DC372755.1 | - | 3 | 252 |
Gene Symbol: NBPF19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003019.1 | XP_016858508.1 | 7 | 853 |
Gene Symbol: NBPF20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278267.1 | NP_001265196.1 | 712 |
Gene Symbol: NBPF25P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104217.1 | 9 | 1037 | |||
GenBank mRNA | AK125792.1 | - | 9 | 1037 |
Gene Symbol: NBPF26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011542529.2 | XP_011540831.1 | 10 | 1423 | ||
XM_017003011.1 | XP_016858500.1 | 10 | 1422 | |||
GenBank mRNA | AK294944.1 | - | 4 | 406 |
Gene Symbol: NBPF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037501.2 | NP_001032590.2 | 10 | 1565 | ||
NR_102404.1 | 10 | 1565 | ||||
NR_102405.1 | 9 | 1462 | ||||
GenBank mRNA | AK307809.1 | - | 8 | 1169 | ||
AY894561.1 | - | 6 | 754 | |||
AY894562.1 | - | 6 | 754 | |||
AY894563.1 | - | 5 | 628 | |||
AY894567.1 | - | 9 | 1462 | |||
AY894568.1 | - | 10 | 1360 | |||
AY894569.1 | - | 10 | 1362 | |||
AY894570.1 | - | 6 | 754 | |||
AY894571.1 | - | 9 | 1462 | |||
AY894572.1 | - | 10 | 1565 | |||
AY894573.1 | - | 9 | 1462 | |||
BC086308.1 | - | 10 | 1408 | 5453529 |
Gene Symbol: NBPF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037675.3 | NP_001032764.2 | 11 | 1468 | ||
NM_001277444.1 | NP_001264373.1 | 11 | 1468 | |||
XM_006711317.2 | XP_006711380.1 | 12 | 1696 | |||
XM_017001278.1 | XP_016856767.1 | 12 | 1696 | |||
GenBank mRNA | AF419618.1 | - | 4 | 416 | ||
AK294414.1 | - | 5 | 626 | |||
AY894574.1 | - | 11 | 1470 | |||
BC071995.1 | - | 6 | 652 | |||
BC094841.1 | - | 6 | 652 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
