siRNA Details
Gene Symbol: NBPF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039703.5 | NP_001034792.4 | 1618 | |||
NM_001302371.1 | NP_001289300.1 | 1618 | ||||
GenBank mRNA | AK000726.1 | - | 10, 11 | 2132 | ||
AY894565.1 | - | 11, 12 | 1655 | |||
BC021111.1 | - | 11, 12 | 1662 | |||
BC063799.1 | - | 11, 12 | 1645 |
Gene Symbol: LOC105371303 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003020.1 | XP_016858509.1 | 10, 11 | 1733 |
Gene Symbol: NBPF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC094705.1 | - | 4, 5 | 610 |
Gene Symbol: NBPF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015383.2 | NP_056198.2 | 10, 11 | 1583 | ||
XM_011509395.2 | XP_011507697.1 | 13, 14 | 2083 | |||
GenBank mRNA | AF379631.1 | - | 1, 2 | 68 | ||
AL117237.1 | - | 4, 5 | 580 |
Gene Symbol: NBPF15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170755.2 | NP_001164226.1 | 12, 13 | 1932 | ||
NM_173638.4 | NP_775909.2 | 9, 10 | 1402 | |||
XM_017001077.1 | XP_016856566.1 | 7, 8 | 1050 | |||
GenBank mRNA | AF379630.1 | - | 4, 5 | 580 | ||
AK097180.1 | - | 12, 13 | 1851 | |||
AK290142.1 | - | 1, 2 | 104 | |||
AK290302.1 | - | 6, 7 | 861 | |||
AK299360.1 | - | 3, 4 | 442 | |||
AY894566.1 | - | 6, 7 | 842 | |||
BC023087.1 | - | 9, 10 | 1259 | 3918932 | ||
DC372755.1 | - | 3, 4 | 442 |
Gene Symbol: NBPF19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003019.1 | XP_016858508.1 | 7, 8 | 1043 |
Gene Symbol: NBPF20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278267.1 | NP_001265196.1 | 902 |
Gene Symbol: NBPF26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011542529.2 | XP_011540831.1 | 10, 11 | 1613 | ||
XM_017003011.1 | XP_016858500.1 | 10, 11 | 1612 | |||
GenBank mRNA | AK294944.1 | - | 4, 5 | 596 |
Gene Symbol: NBPF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037501.2 | NP_001032590.2 | 10, 11 | 1755 | ||
NR_102404.1 | 10, 11 | 1755 | ||||
NR_102405.1 | 9, 10 | 1652 | ||||
GenBank mRNA | AK300749.1 | - | 1, 2 | 15 | ||
AK307809.1 | - | 8, 9 | 1359 | |||
AY894561.1 | - | 6, 7 | 944 | |||
AY894562.1 | - | 6, 7 | 944 | |||
AY894563.1 | - | 5, 6 | 818 | |||
AY894567.1 | - | 9, 10 | 1652 | |||
AY894568.1 | - | 10, 11 | 1550 | |||
AY894569.1 | - | 10, 11 | 1552 | |||
AY894570.1 | - | 6, 7 | 944 | |||
AY894572.1 | - | 10, 11 | 1755 | |||
BC086308.1 | - | 10, 11 | 1598 | 5453529 |
Gene Symbol: NBPF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037675.3 | NP_001032764.2 | 11, 12 | 1658 | ||
NM_001277444.1 | NP_001264373.1 | 11, 12 | 1658 | |||
XM_006711317.2 | XP_006711380.1 | 12, 13 | 1886 | |||
XM_017001278.1 | XP_016856767.1 | 12, 13 | 1886 | |||
GenBank mRNA | AF419618.1 | - | 4, 5 | 606 | ||
AK294414.1 | - | 5, 6 | 816 | |||
AY894574.1 | - | 11, 12 | 1660 | |||
BC071995.1 | - | 6, 7 | 842 | |||
BC094841.1 | - | 6, 7 | 842 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
