siRNA Details
Gene Symbol: ADAMTSL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145320.1 | NP_001138792.1 | 13 | 2398 | ||
NM_014694.3 | NP_055509.2 | 13 | 2056 | |||
XM_005272237.3 | XP_005272294.1 | 13 | 2253 | |||
XM_005272238.3 | XP_005272295.1 | 12 | 1895 | |||
XM_005272239.2 | XP_005272296.1 | 13 | 1931 | |||
XM_011519241.2 | XP_011517543.2 | 12 | 2074 | |||
XM_011519242.2 | XP_011517544.1 | 12 | 1930 | |||
GenBank mRNA | AB011177.1 | - | 13 | 2398 | ||
AK127107.1 | - | 1 | 1752 | |||
AK293498.1 | - | 3 | 426 | |||
BC050544.1 | - | 13 | 2056 | 6196393 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
