siRNA Details
Gene Symbol: QRFP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198180.2 | NP_937823.1 | 3 | 708 | ||
GenBank mRNA | AB109625.1 | - | 1 | 187 | ||
BC101127.2 | - | 1 | 187 | 40014164 | ||
BC101128.1 | - | 1 | 187 | 40014167 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
