siRNA Details
Gene Symbol: CPTP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029885.1 | NP_001025056.1 | 3 | 923 | ||
XM_005244801.3 | XP_005244858.1 | 3 | 1036 | |||
XM_005244802.1 | XP_005244859.1 | 2 | 733 | |||
XM_011542200.2 | XP_011540502.1 | 3 | 932 | |||
GenBank mRNA | AL122073.1 | - | 1 | 31 | ||
BC098429.1 | - | 1 | 347 | |||
JN542538.1 | - | 2 | 508 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
