siRNA Details
Gene Symbol: SIRT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016538.2 | NP_057622.1 | 5 | 502 | ||
XM_006722284.2 | XP_006722347.1 | 3 | 236 | |||
XM_011523580.1 | XP_011521882.1 | 4 | 368 | |||
XM_011523581.2 | XP_011521883.1 | 4 | 2105 | |||
GenBank mRNA | AF233395.1 | - | 5 | 484 | ||
AK002027.1 | - | 5 | 472 | |||
AK094326.1 | - | 4 | 567 | |||
AK131437.1 | - | 2 | 1585 | |||
AK131484.1 | - | 1 | 984 | |||
AK290265.1 | - | 5 | 470 | |||
AL137626.1 | - | 2 | 70 | |||
BC017305.2 | - | 5 | 466 | 5087554 | ||
BC101791.1 | - | 5 | 483 | 8069297 | ||
BC101793.1 | - | 5 | 482 | 8069299 | ||
BQ890203.1 | - | 5 | 502 | |||
CD579273.2 | - | 5 | 488 |

More Information
Other Species Targeted

Gene Ontology Categories:
