siRNA Details
Gene Symbol: SIRT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016538.2 | NP_057622.1 | 7 | 761 | ||
XM_006722284.2 | XP_006722347.1 | 5 | 495 | |||
XM_011523580.1 | XP_011521882.1 | 6 | 627 | |||
XM_011523581.2 | XP_011521883.1 | 6 | 2364 | |||
GenBank mRNA | AF233395.1 | - | 7 | 743 | ||
AK002027.1 | - | 5 | 1541 | |||
AK094326.1 | - | 6 | 826 | |||
AK131437.1 | - | 4 | 1818 | |||
AK131484.1 | - | 2 | 1316 | |||
AK290265.1 | - | 7 | 729 | |||
AL137626.1 | - | 4 | 329 | |||
BC017305.2 | - | 7 | 725 | 5087554 | ||
BC101791.1 | - | 7 | 742 | 8069297 | ||
BC101793.1 | - | 7 | 741 | 8069299 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
