siRNA Details
Gene Symbol: SIRT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016538.2 | NP_057622.1 | 7 | 835 | ||
XM_006722284.2 | XP_006722347.1 | 5 | 569 | |||
XM_011523580.1 | XP_011521882.1 | 6 | 701 | |||
XM_011523581.2 | XP_011521883.1 | 6 | 2438 | |||
GenBank mRNA | AF233395.1 | - | 7 | 817 | ||
AK002027.1 | - | 5 | 1615 | |||
AK094326.1 | - | 6 | 900 | |||
AK131437.1 | - | 4 | 1892 | |||
AK131484.1 | - | 2 | 1390 | |||
AK290265.1 | - | 7 | 803 | |||
AL137626.1 | - | 4 | 403 | |||
BC017305.2 | - | 7 | 799 | 5087554 | ||
BC101791.1 | - | 7 | 816 | 8069297 | ||
BC101793.1 | - | 7 | 815 | 8069299 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
