siRNA Details
Gene Symbol: IDO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194294.2 | NP_919270.2 | 2 | 353 | ||
GenBank mRNA | AK128691.1 | - | 2 | 353 | ||
BC113496.1 | - | 2 | 132 | |||
BC113498.1 | - | 2 | 132 | |||
EF052681.1 | - | 1 | 72 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
