siRNA Details
Gene Symbol: HHIPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024746.3 | NP_079022.2 | 4 | 1422 | ||
XM_011509985.2 | XP_011508287.1 | 4 | 1367 | |||
XM_011509986.1 | XP_011508288.1 | 4 | 1367 | |||
XM_017002350.1 | XP_016857839.1 | 4 | 1367 | |||
GenBank mRNA | AK023902.1 | - | 4 | 1107 | ||
AY358602.1 | - | 4 | 1404 | |||
BC007638.1 | - | 4 | 1408 | 3533045 | ||
BC016552.1 | - | 4 | 1422 | 4452939 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
