siRNA Details
Gene Symbol: NKX6-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152568.3 | NP_689781.1 | 1 | 149 | ||
GenBank mRNA | AK057898.1 | - | 1 | 149 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
