siRNA Details
Gene Symbol: PKD1P6-NPIPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_123721.1 | 18 | 3887 | |||
NR_123722.1 | 8 | 1748 | ||||
GenBank mRNA | AK023376.1 | - | 10 | 2332 | ||
AK131084.1 | - | 13 | 4251 | |||
AK131085.1 | - | 12 | 4157 | |||
AK302708.1 | - | 8 | 1748 | |||
AK303260.1 | - | 8 | 1261 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
