siRNA Details
Gene Symbol: PKD1P6-NPIPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_123721.1 | 14 | 3159 | |||
NR_123722.1 | 4 | 1020 | ||||
GenBank mRNA | AK023376.1 | - | 6 | 1604 | ||
AK131084.1 | - | 9 | 3523 | |||
AK131085.1 | - | 8 | 3424 | |||
AK302708.1 | - | 4 | 1020 | |||
AK303260.1 | - | 4 | 533 |
Gene Symbol: NPIPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK160377.1 | - | 14 | 3172 |
Gene Symbol: PKD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000296.3 | NP_000287.3 | 26 | 9505 | ||
NM_001009944.2 | NP_001009944.2 | 26 | 9505 | |||
XM_005255370.3 | XP_005255427.1 | 16 | 7092 | |||
XM_011522528.2 | XP_011520830.1 | 27 | 9530 | |||
XM_011522529.2 | XP_011520831.1 | 27 | 9531 | |||
XM_011522530.2 | XP_011520832.1 | 27 | 9335 | |||
XM_011522532.2 | XP_011520834.1 | 26 | 9263 | |||
XM_011522534.2 | XP_011520836.1 | 27 | 9320 | |||
XM_011522535.2 | XP_011520837.1 | 18 | 7552 | |||
XM_011522537.2 | XP_011520839.1 | 16 | 6463 | |||
XM_017023285.1 | XP_016878774.1 | 17 | 7107 | |||
GenBank mRNA | AB209025.1 | - | 13 | 3859 | ||
AB209675.1 | - | 12 | 4773 | |||
L33243.1 | - | 26 | 9507 | |||
U24497.1 | - | 26 | 9431 |
Gene Symbol: PKD1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036447.1 | 20 | 4290 | |||
GenBank mRNA | AK124516.1 | - | 15 | 2545 | ||
AK126798.1 | - | 15 | 2457 | |||
AK304604.1 | - | 1 | 76 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
