siRNA Details
Gene Symbol: ZNF322 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242797.1 | NP_001229726.1 | 5 | 933 | ||
NM_001242798.1 | NP_001229727.1 | 3 | 788 | |||
NM_001242799.1 | NP_001229728.1 | 3 | 769 | |||
NM_024639.4 | NP_078915.2 | 4 | 858 | |||
GenBank mRNA | AK290038.1 | - | 4 | 583 | ||
AY376736.1 | - | 4 | 641 | |||
BC050425.1 | - | 1 | 354 | 6473688 | ||
BC074772.2 | - | 1 | 150 | |||
BC119669.2 | - | 1 | 271 | 40116184 |
Gene Symbol: ZNF214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013249.2 | NP_037381.2 | 3 | 1628 | ||
XM_005253128.3 | XP_005253185.1 | 4 | 1703 | |||
XM_005253129.3 | XP_005253186.1 | 3 | 1636 | |||
XM_006718308.3 | XP_006718371.1 | 3 | 1636 | |||
XM_006718309.3 | XP_006718372.1 | 3 | 1632 | |||
XM_011520355.2 | XP_011518657.1 | 4 | 1699 | |||
XM_011520357.2 | XP_011518659.1 | 3 | 1390 | |||
XM_017018269.1 | XP_016873758.1 | 3 | 1404 | |||
GenBank mRNA | AF056617.1 | - | 3 | 1583 | ||
BC136772.1 | - | 3 | 1508 | 9020762 | ||
BC144405.1 | - | 4 | 1567 | 9052926 |

More Information
Other Species Targeted

Gene Ontology Categories:
