siRNA Details
Gene Symbol: CCDC34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030771.1 | NP_110398.1 | 2 | 428 | ||
NM_080654.2 | NP_542385.1 | 2 | 428 | |||
GenBank mRNA | AF155108.1 | - | 2 | 1054 | ||
AF382034.1 | - | 2 | 317 | |||
AK308494.1 | - | 3 | 376 | |||
AK313358.1 | - | 2 | 395 | |||
AL555720.3 | - | 2 | 412 | |||
AY598334.1 | - | 2 | 374 | |||
BC008496.1 | - | 2 | 396 | 4276656 | ||
BC029421.1 | - | 2 | 103 | |||
BU595712.1 | - | 431 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
