siRNA Details
Gene Symbol: CCDC34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030771.1 | NP_110398.1 | 3 | 593 | ||
NM_080654.2 | NP_542385.1 | 3 | 593 | |||
GenBank mRNA | AF155108.1 | - | 3 | 1219 | ||
AF382034.1 | - | 3 | 481 | |||
AK308494.1 | - | 4 | 541 | |||
AK313358.1 | - | 3 | 560 | |||
AL555720.3 | - | 3 | 577 | |||
AY598334.1 | - | 3 | 539 | |||
BC008496.1 | - | 3 | 561 | 4276656 | ||
BC029421.1 | - | 3 | 268 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
