siRNA Details
Gene Symbol: CPTP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029885.1 | NP_001025056.1 | 3 | 759 | ||
XM_005244801.3 | XP_005244858.1 | 3 | 872 | |||
XM_005244802.1 | XP_005244859.1 | 2 | 569 | |||
XM_011542200.2 | XP_011540502.1 | 3 | 768 | |||
GenBank mRNA | BC098429.1 | - | 1 | 183 | ||
JN542538.1 | - | 2 | 344 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
