siRNA Details
Gene Symbol: PDGFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002607.5 | NP_002598.4 | 3 | 1096 | ||
NM_033023.4 | NP_148983.1 | 3 | 1096 | |||
XM_011515415.1 | XP_011513717.1 | 4 | 588 | |||
XM_011515416.1 | XP_011513718.1 | 4 | 592 | |||
XM_011515417.1 | XP_011513719.1 | 4 | 588 | |||
XM_011515418.1 | XP_011513720.1 | 4 | 592 | |||
XM_011515419.2 | XP_011513721.1 | 3 | 296 | |||
XM_017012289.1 | XP_016867778.1 | 3 | 329 | |||
GenBank mRNA | A09204.1 | - | 3 | 289 | ||
AB451309.1 | - | 3 | 253 | |||
AB451439.1 | - | 3 | 253 | |||
AU118732.1 | - | 3 | 424 | |||
BC109246.1 | - | 1 | 1116 | |||
BE336646.1 | - | 2 | 210 | |||
X03795.1 | - | 3 | 640 | |||
X06374.1 | - | 3 | 656 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
