siRNA Details
Gene Symbol: POTEC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137671.1 | NP_001131143.1 | 5 | 1427 | ||
NR_134878.1 | 5 | 1427 | ||||
GenBank mRNA | AK307658.1 | - | 5 | 1427 | ||
BC140940.1 | - | 5 | 1320 | 9056752 | ||
BX649118.1 | - | 5 | 1413 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
