siRNA Details
Gene Symbol: POTEE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083538.1 | NP_001077007.1 | 2 | 642 | ||
XM_011511216.2 | XP_011509518.1 | 2 | 720 | |||
XM_017004159.1 | XP_016859648.1 | 2 | 721 | |||
XM_017004161.1 | XP_016859650.1 | 4 | 1079 | |||
GenBank mRNA | AY462868.1 | - | 2 | 642 | ||
AY462871.1 | - | 2 | 642 | |||
AY462872.1 | - | 2 | 642 | |||
AY462873.1 | - | 2 | 642 |
Gene Symbol: LOC100288966 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257362.1 | NP_001244291.1 | 2 | 642 | ||
XM_006709930.3 | XP_006709993.1 | 2 | 738 | |||
XM_011545655.2 | XP_011543957.1 | 2 | 738 | |||
XM_011545656.2 | XP_011543958.1 | 2 | 738 | |||
GenBank mRNA | AY172978.1 | - | 2 | 642 |
Gene Symbol: POTEC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137671.1 | NP_001131143.1 | 2 | 1044 | ||
NR_134878.1 | 2 | 1044 | ||||
GenBank mRNA | AK307658.1 | - | 2 | 1044 | ||
BC140940.1 | - | 2 | 937 | 9056752 | ||
BX649118.1 | - | 2 | 1030 |
Gene Symbol: POTED | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174981.3 | NP_778146.2 | 2 | 642 | ||
XM_006723997.3 | XP_006724060.1 | 2 | 738 | |||
XM_011529550.2 | XP_011527852.1 | 2 | 738 |
Gene Symbol: POTEF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099771.2 | NP_001093241.1 | 4 | 990 | ||
XM_017004833.1 | XP_016860322.1 | 2 | 735 | |||
GenBank mRNA | EF523384.1 | - | 4 | 992 |
Gene Symbol: POTEG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005356.2 | NP_001005356.1 | 2 | 642 | ||
NR_027480.1 | 2 | 642 | ||||
GenBank mRNA | AY462874.1 | - | 2 | 642 | ||
AY465171.1 | - | 3 | 642 | |||
AY465172.1 | - | 3 | 642 | |||
BC127623.1 | - | 2 | 622 |
Gene Symbol: POTEH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136213.1 | NP_001129685.1 | 2 | 753 | ||
GenBank mRNA | AY466021.1 | - | 2 | 753 |
Gene Symbol: POTEI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277406.1 | NP_001264335.1 | 2 | 590 | ||
XM_011511670.2 | XP_011509972.1 | 2 | 732 | |||
XM_017004732.1 | XP_016860221.1 | 3 | 711 |
Gene Symbol: POTEJ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277083.1 | NP_001264012.1 | 2 | 479 | ||
XM_011511683.2 | XP_011509985.1 | 2 | 621 | |||
XM_017004741.1 | XP_016860230.1 | 3 | 594 |
Gene Symbol: POTEKP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033885.2 | 1 | 81 | |||
GenBank mRNA | BM763453.1 | - | 1 | 81 |
Gene Symbol: POTEM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145442.1 | NP_001138914.1 | 2 | 642 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
