siRNA Details
Gene Symbol: POTEB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207355.3 | NP_997238.2 | 4 | 1220 | ||
NR_110752.1 | 4 | 1220 | ||||
XM_006720358.2 | XP_006720421.1 | 4 | 1267 | |||
XM_011543795.2 | XP_011542097.1 | 4 | 1267 | |||
XM_011543796.2 | XP_011542098.1 | 4 | 1267 | |||
XM_011543797.2 | XP_011542099.1 | 4 | 1267 | |||
GenBank mRNA | AY465170.1 | - | 5 | 790 | ||
AY465173.1 | - | 4 | 897 | |||
BC101005.1 | - | 4 | 771 | 40006483 | ||
BC101006.1 | - | 4 | 882 | 40006485 | ||
BC127123.1 | - | 4 | 882 | 40006490 |
Gene Symbol: LOC100288966 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257362.1 | NP_001244291.1 | 4 | 901 | ||
XM_006709930.3 | XP_006709993.1 | 4 | 997 | |||
XM_011545655.2 | XP_011543957.1 | 4 | 997 | |||
XM_011545656.2 | XP_011543958.1 | 4 | 997 | |||
GenBank mRNA | AY172978.1 | - | 4 | 901 |
Gene Symbol: POTEC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137671.1 | NP_001131143.1 | 4 | 1303 | ||
NR_134878.1 | 4 | 1303 | ||||
GenBank mRNA | AK307658.1 | - | 4 | 1303 | ||
BC140940.1 | - | 4 | 1196 | 9056752 | ||
BX649118.1 | - | 4 | 1289 |
Gene Symbol: POTED | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174981.3 | NP_778146.2 | 4 | 901 | ||
XM_006723997.3 | XP_006724060.1 | 4 | 997 | |||
XM_011529550.2 | XP_011527852.1 | 4 | 997 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
