siRNA Details
Gene Symbol: CCL4L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207007.3 | NP_996890.1 | 2 | 379 | ||
GenBank mRNA | AY766447.1 | - | 2 | 224 | ||
BC070310.1 | - | 2 | 221 | 6708338 | ||
BC130456.1 | - | 2 | 186 | 40146487 | ||
BC130458.1 | - | 2 | 186 | 40146489 | ||
BC144393.1 | - | 2 | 186 | 9052914 | ||
BC144394.1 | - | 2 | 186 | 9052915 | ||
BC146944.1 | - | 2 | 379 | 9056872 | ||
BC146958.1 | - | 2 | 379 | 9056886 | ||
BC171857.1 | - | 2 | 379 | 9054511 | ||
BC171864.1 | - | 2 | 379 | 9054518 |
Gene Symbol: CCL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002984.3 | NP_002975.1 | 2 | 224 | ||
GenBank mRNA | AA362169.1 | - | 2 | 201 | ||
AY766446.1 | - | 2 | 224 | |||
BC104226.1 | - | 2 | 215 | 40032168 | ||
BC104227.1 | - | 2 | 215 | 40032172 | ||
BC107433.1 | - | 2 | 225 | 6727378 | ||
CR542119.1 | - | 2 | 145 | |||
J04130.1 | - | 2 | 253 | |||
M25316.1 | - | 2 | 218 | |||
M57503.1 | - | 2 | 132 | |||
X16166.1 | - | 2 | 197 | |||
X53683.1 | - | 2 | 211 |
Gene Symbol: CCL4L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291468.1 | NP_001278397.1 | 2 | 224 | ||
NM_001291469.1 | NP_001278398.1 | 2 | 224 | |||
NM_001291470.1 | NP_001278399.1 | 2 | 224 | |||
NM_001291471.1 | NP_001278400.1 | 2 | 224 | |||
NM_001291472.1 | NP_001278401.1 | 2 | 224 | |||
NM_001291473.1 | NP_001278402.1 | 2 | 224 | |||
NM_001291475.1 | NP_001278404.1 | 2 | 224 | |||
GenBank mRNA | AY766450.1 | - | 2 | 224 | ||
AY766451.1 | - | 2 | 224 | |||
AY766452.1 | - | 2 | 224 | |||
AY766453.1 | - | 2 | 224 | |||
AY766454.1 | - | 2 | 224 | |||
AY766455.1 | - | 2 | 224 | |||
AY766458.1 | - | 2 | 224 | |||
BC092445.1 | - | 2 | 221 | 30416435 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
