siRNA Details
Gene Symbol: CCL4L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291468.1 | NP_001278397.1 | 3 | 491 | ||
NM_001291469.1 | NP_001278398.1 | 3 | 695 | |||
NM_001291470.1 | NP_001278399.1 | 2 | 1097 | |||
NM_001291471.1 | NP_001278400.1 | 3 | 675 | |||
NM_001291472.1 | NP_001278401.1 | 3 | 654 | |||
NM_001291473.1 | NP_001278402.1 | 3 | 624 | |||
NM_001291474.1 | NP_001278403.1 | 2 | 580 | |||
NM_001291475.1 | NP_001278404.1 | 3 | 685 | |||
NR_111970.1 | 2 | 376 | ||||
GenBank mRNA | AY766450.1 | - | 3 | 491 | ||
AY766451.1 | - | 3 | 695 | |||
AY766452.1 | - | 2 | 1097 | |||
AY766453.1 | - | 3 | 675 | |||
AY766454.1 | - | 3 | 654 | |||
AY766455.1 | - | 3 | 624 | |||
AY766456.1 | - | 2 | 376 | |||
AY766457.1 | - | 2 | 580 | |||
AY766458.1 | - | 3 | 685 | |||
BC092445.1 | - | 3 | 692 | 30416435 | ||
CD521054.1 | - | 1 | 424 |
Gene Symbol: CCL4L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207007.3 | NP_996890.1 | 3 | 661 | ||
NR_111969.1 | 2 | 546 | ||||
GenBank mRNA | AY766447.1 | - | 3 | 506 | ||
AY766449.1 | - | 2 | 391 | |||
BC070310.1 | - | 3 | 503 | 6708338 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
