siRNA Details
Gene Symbol: HSF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005526.2 | NP_005517.1 | 4 | 549 | ||
XM_005272315.2 | XP_005272372.1 | 4 | 549 | |||
XM_005272316.2 | XP_005272373.1 | 4 | 549 | |||
XM_005272317.1 | XP_005272374.1 | 4 | 549 | |||
XM_011517004.2 | XP_011515306.1 | 2 | 150 | |||
XM_017013377.1 | XP_016868866.1 | 4 | 549 | |||
GenBank mRNA | AK125467.1 | - | 2 | 574 | ||
AK222497.1 | - | 4 | 533 | |||
AK290975.1 | - | 4 | 529 | |||
AK300205.1 | - | 1 | 31 | |||
BC014638.1 | - | 4 | 520 | 3528599 | ||
BT007351.1 | - | 4 | 379 | |||
DA481620.1 | - | 4 | 549 | |||
M64673.1 | - | 4 | 539 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
