siRNA Details
Gene Symbol: HSF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005526.2 | NP_005517.1 | 10 | 1401 | ||
XM_005272315.2 | XP_005272372.1 | 11 | 1485 | |||
XM_005272316.2 | XP_005272373.1 | 11 | 1485 | |||
XM_011517004.2 | XP_011515306.1 | 8 | 1010 | |||
XM_011517006.2 | XP_011515308.1 | 6 | 792 | |||
XM_017013377.1 | XP_016868866.1 | 10 | 1401 | |||
GenBank mRNA | AF299094.1 | - | 2 | 121 | ||
AK054762.1 | - | 6 | 1518 | |||
AK125467.1 | - | 7 | 2703 | |||
AK126580.1 | - | 1 | 987 | |||
AK222497.1 | - | 10 | 1385 | |||
AK290975.1 | - | 10 | 1381 | |||
AK300205.1 | - | 7 | 883 | |||
BC014638.1 | - | 10 | 1372 | 3528599 | ||
BT007351.1 | - | 10 | 1231 | |||
M64673.1 | - | 10 | 1391 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
