siRNA Details
Gene Symbol: CRB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139161.4 | NP_631900.1 | 3 | 315 | ||
NM_174881.3 | NP_777377.1 | 3 | 315 | |||
NM_174882.2 | NP_777378.1 | 3 | 639 | |||
XM_017027471.1 | XP_016882960.1 | 2 | 327 | |||
GenBank mRNA | AF503290.1 | - | 3 | 274 | ||
AK000653.1 | - | 3 | 254 | |||
AK292956.1 | - | 3 | 280 | |||
AY103469.1 | - | 3 | 640 | |||
AY358684.1 | - | 3 | 270 | |||
BC002652.2 | - | 2 | 141 | 3610257 | ||
BC018409.1 | - | 3 | 266 | 3861124 | ||
BM129809.1 | - | 3 | 283 | |||
DB215367.1 | - | 3 | 316 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
