siRNA Details
Gene Symbol: PABPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002568.3 | NP_002559.2 | 14 | 2352 | ||
XM_005250861.2 | XP_005250918.1 | 14 | 3263 | |||
GenBank mRNA | AK095194.1 | - | 14 | 2187 | ||
AK130523.1 | - | 14 | 2352 | |||
AK298990.1 | - | 15 | 2068 | |||
AK303120.1 | - | 15 | 2076 | |||
BC015958.2 | - | 15 | 2247 | 3940309 | ||
BC023217.1 | - | 14 | 2330 | |||
BC023520.2 | - | 14 | 2331 | 4123269 | ||
BC041863.1 | - | 14 | 2976 | |||
Y00345.1 | - | 14 | 2341 | |||
Z48501.1 | - | 12 | 1506 |
Gene Symbol: PABPC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030979.2 | NP_112241.2 | 1 | 1894 | ||
GenBank mRNA | AF132026.1 | - | 1 | 1856 | ||
AK313336.1 | - | 1 | 1876 | |||
AL136900.1 | - | 1 | 1876 | |||
BC027617.2 | - | 1 | 1860 | 4829121 | ||
BC045608.1 | - | 1 | 1870 | 5273503 |
Gene Symbol: RLIM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016120.3 | NP_057204.2 | 4 | 10392 | ||
NM_183353.2 | NP_899196.1 | 5 | 10454 | |||
GenBank mRNA | AK127880.1 | - | 1 | 2241 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
